Z278327
Conical reaction vial
capacity 10 mL
About This Item
Código UNSPSC:
41121700
NACRES:
NB.43
Produtos recomendados
Materiais
colorless tube
glass tube
screw top neck
esterilidade
non-sterile
Características
graduations: no
External thread
embalagem
pack of 1 ea
fabricante/nome comercial
Ace Glass D104153
capacidade
10 mL
Categorias relacionadas
Descrição geral
Thin-wall with threaded outer 14/10 joint, cap with hole, PTFE-faced silicone rubber septum, and FETFE® O-ring.
The product does NOT have the side arm opening. It is the product shown on the left of the photo.
The product does NOT have the side arm opening. It is the product shown on the left of the photo.
Informações legais
FETFE is a registered trademark of Ace Glass, Inc.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z278327-1EA | 4061832349367 |