Z268143
Aldrich® desiccator cabinet
H × W × D 9 in. × 9 in. × 16 in., 2 trays, clear
About This Item
Código UNSPSC:
41104510
NACRES:
NB.43
Produtos recomendados
A × L × P
9 in. × 9 in. × 16 in.
cor
clear
Descrição geral
Aldrich desiccator cabinets are dust and moisture-free storage for refrigerators, freezers, and at room temperature. The clear walls, top, and base are 1/4 in. thick in acrylic for 360° visability.
Gasketed, air-tight doors are 3/8 in. thick acrylic with full length hinges and SS twist locking latches. Clear acrylic, deep slide-in trays with perforated bottoms for proper circulation within the cabinet. Dark bronze acrylic provides maximum protection for light-sensitive materials.
Gasketed, air-tight doors are 3/8 in. thick acrylic with full length hinges and SS twist locking latches. Clear acrylic, deep slide-in trays with perforated bottoms for proper circulation within the cabinet. Dark bronze acrylic provides maximum protection for light-sensitive materials.
Informações legais
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Os clientes também visualizaram
Global Trade Item Number
SKU | GTIN |
---|---|
Z268143-1EA | 4061837618086 |