Z173118
Aldrich® Schlenk-type adapter
Straight with glass stopcock on side-arm, male joint: threaded 14/20, female joint: threaded 14/20
About This Item
Código UNSPSC:
41121815
NACRES:
NB.43
Produtos recomendados
Junta
female joint: threaded14/20
male joint: threaded14/20
Descrição geral
Thermoplastic screw caps with Viton O-rings usable to 140 deg C.
Ligação
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Informações legais
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z173118-1EA | 4061833303658 |