Documentos Principais
Z143944
Solid-glass beads
borosilicate, diam. 5 mm
Sinônimo(s):
borosilicate beads
About This Item
Código UNSPSC:
41102422
NACRES:
NB.43
Materiais
borosilicate
embalagem
pack of 1 ea
pack of 4 ea
Diâmetro
5 mm
Categorias relacionadas
Descrição geral
For use as distillation column packing, mixing beads, and boiling stones. Each 454 g box has a packing volume of approx. 360 cm3. +/-10% variation in bead size.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Visite a Biblioteca de Documentos
Os clientes também visualizaram
Slide 1 of 9
1 of 9
Glass beads diam. 0.5 mm, for Mini-Beadbeater
Sigma-Aldrich
Z250465
Glass beads
Solid-glass beads borosilicate, diam. 2 mm
Z273627
Solid-glass beads
Glass beads 6 mm
Supelco
1.04018
Glass beads 6 mm
Glass beads diam. 1.0 mm, for Beadbeating
Sigma-Aldrich
Z250473
Glass beads
Pyrex® solid glass beads 3mm packing beads, 4/cs
CLS72683
Pyrex® solid glass beads
Pyrex® solid glass beads 4mm packing beads, 4/cs
CLS72684
Pyrex® solid glass beads
Global Trade Item Number
SKUGTIN
Z143944-4EA4061833576809
Z143944-1EA4061833576793