Z104485
Tubing quick-disconnects
for tubing i.d., 1/8 ‑ 1/4 in.
About This Item
Código UNSPSC:
41121815
NACRES:
NB.35
Produtos recomendados
Materiais
PE
Características
for tubing i.d. 1/8 ‑ 1/4 in.
embalagem
pack of 12 ea
fabricante/nome comercial
Bel-Art H19727-0000
Categorias relacionadas
Descrição geral
Consists of two serrated connectors which are joined securely through a male-female center taper. Connector of one size can be interchanged with other sizes to produce reducing combinations.
- Easy-to-assemble barbed connectors require no tools
- Ideal for use with flexible tubing
- Handy on lines that require frequent disconnection
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Os clientes também visualizaram
Global Trade Item Number
SKU | GTIN |
---|---|
Z104485-12EA | 4061832261515 |