TEM-G300F1-AU
Gilder Finder Grids G300F1
Gold, size 300 mesh, vial of 100
About This Item
Código UNSPSC:
41111761
NACRES:
MA.24
Produtos recomendados
embalagem
vial of 100
tamanho
300 mesh
Descrição geral
Gilder Finder Grids G300F1. Gold grids, 300 mesh. The block of 625 (25x25) individual cells is divided into smaller blocks bounded by thicker crenellated bars. Each small block of 25 cells has a central alphabet letter from A to Y for identification purposes.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
TEM-G300F1-AU | 4065266052015 |