CAD9881
Dissolution cannulae
gauge 14, 316 stainless steel hub, L 7 1/2 in.
About This Item
Código UNSPSC:
41122002
NACRES:
NB.03
Produtos recomendados
Materiais
316 stainless steel hub
embalagem
pack of 1 ea
fabricante/nome comercial
Cadence Science 9881
Comprimento
7 1/2 in.
medidor
14
Categorias relacionadas
Descrição geral
Ideal for drug dissolution testing. Drug dissolution testing is routinely used in the pharmaceutical industry to provide critical release information for quality control purposes, in new drug development, and to predict in vivo drug release profiles. These cannula feature 316 stainless steel hub and tubing for maximum resistance to chemical reaction.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Os clientes também visualizaram
Global Trade Item Number
SKU | GTIN |
---|---|
CAD9881-1EA | 4061832518541 |