CAD4071
Standard point hypodermic needles, 304 stainless steel
gauge 18, L 1 1/2 in.
About This Item
Código UNSPSC:
42142523
NACRES:
NB.03
Produtos recomendados
Materiais
304 stainless steel needle
embalagem
pack of 12 ea
fabricante/nome comercial
Cadence Science 4071
Comprimento
1 1/2 in.
medidor
18
Categorias relacionadas
Descrição geral
Hypodermic needles are precision made and designed to ensure strict adherence with U.S. Government specification GG-N-196. Points are machine ground to achieve uniform sharpness. Precision grinding, micro-bead jet blasting, and ultrasonic cleaning assure freedom from burrs and debris. All needle cannula are fabricated of type 304 stainless steel tubing which must pass rigorous quality control prior to acceptance and use.
Standard nickle plated Luer hubs comply with Luer taper specifications ISO 594. This assures an interchangeable fit mating with Luer slips and Luer locks. Contoured hub cavity facilitates cleaning.
Standard nickle plated Luer hubs comply with Luer taper specifications ISO 594. This assures an interchangeable fit mating with Luer slips and Luer locks. Contoured hub cavity facilitates cleaning.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
CAD4071-12EA | 4061832516684 |