BAF420731009
SP Bel-Art® Secador® 3.0 Gas-Purge Desiccator Cabinet
volume 1.6 ft3
About This Item
Código UNSPSC:
41123002
NACRES:
NB.77
Produtos recomendados
Materiais
amber
polycarbonate
Características
Can be stacked three high
embalagem
pkg of 1 ea
fabricante/nome comercial
Bel-Art F42073-1009
L × P
34.1 cm (13.4 in.) × 41.4 cm (16.3 in.)
volume
1.6 ft3
Descrição geral
The attractive design and sturdy construction of the Secador® line of desiccator cabinets assures easy and reliable storage of moisture and UV sensitive products. Choose from an extensive selection of sizes, styles and colors to meet your specific dry storage needs. U.S. Patents: 6,834,920; 7,318,630 and D474,626
- Polycarbonate construction blocks 99% of UV light* and is resistant to staining, crazing and chemical attack
- Rear mounted gas ports are tapered to accept tubing with ¼ to ⅜” I.D.
- Amber color models reduce visible light penetration by over 50% making it the perfect choice for storing light sensitive materials like reagents, analytical standards and investigational compounds
- Large doors maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
- Perforated shelves facilitate circulation of dry air; space for desiccant under bottom shelf
- Easy-to-view digital hygrometer is located in the door front ; side swinging door opens to the left
- Compact 34.1W x 41.4cmD footprint for space saving and can be stacked up to 3 units high
- Replacement shelves available; 4.0 units can be converted from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves.
- Gas ports can be closed to use desiccator with standard desiccant cartridges
Informações legais
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Secador is a registered trademark of SP Scienceware, a division of SP
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BAF420731009-1EA | 4065269239796 |