Merck
Z719528
Sorenson low binding Ultra G microtips
maximum volume 10 μL, pack of 96 ea CellRack, sterile
About This Item
UNSPSC-Code:
41121600
NACRES:
NB.01
Materialien
colorless
polypropylene
Sterilität
sterile
Leistungsmerkmale
barrier: no
graduations
Verpackung
case of 4800 ea 50 QuickRack of 96 tips
pack of 96 ea CellRack
pack of 960 ea 10 QuickRack of 96 tips
Hersteller/Markenname
Sorenson 23570T
Maximales Volumen
10 μL
Volumen
10 μL
Allgemeine Beschreibung
Tips feature a new surface technology designed for pipetting substances which tend to stick to the walls of pipette tips or for pipetting expensive reagents. Universal fit for ultra-micro pipettors.
Rechtliche Hinweise
Sorenson is a trademark of Sorenson BioScience, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)