Merck
TMO4112-2000
Nalgene® sterile single-use Erlenmeyer flask with solid closure
PETG, bottle volume 2000 mL, case of 4 ea
Synonym(e):
Nalgene Erlenmeyer flask, single use plastic flask
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.13
Materialien
PETG
Sterilität
sterile
Verpackung
case of 4 ea
Hersteller/Markenname
Nalgene 4112-2000
Flaschenvolumen
2000 mL
Allgemeine Beschreibung
Save preparation time and avoid contamination risk by using Thermo Scientific Nalgene Single-Use PETG Erlenmeyer Flasks with Baffled Bottom, ideal for suspension of cell culture, media preparation, mixing and storage. Crystal-clear PETG is robust and break resistant, increasing personal safety. All sizes are available vented or non-vented.
Sonstige Hinweise
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)