OH30378544
Ohaus® Dissolved Oxygen Electrode
STDO21, cable L 1 m
Alle Fotos(1)
About This Item
UNSPSC-Code:
39121436
Empfohlene Produkte
Verpackung
pack of 1 ea
Hersteller/Markenname
Ohaus® 30378544
dissolved oxygen measuring range
0.00-20.0 mg/L
Kabel L
1 m
Allgemeine Beschreibung
All electrodes include a temperature sensor, which powers automatic temperature compensation and ensures accurate measurements.
Anwendung
pH, reference, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO), temperature measurement
Leistungsmerkmale und Vorteile
Can be used in conjunction with all Starter bench and portable meters
Rechtliche Hinweise
Ohaus is a registered trademark of Ohaus Corp.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)