OH30073981
Ohaus® Pen pH Meter
Model ST20T-B
Alle Fotos(1)
About This Item
UNSPSC-Code:
41115603
Empfohlene Produkte
Materialien
epoxy shaft
Verpackung
pack of 1 ea
Hersteller/Markenname
Ohaus® 30073981
Messbereich TDS
0.0-100.0 mg/L
Temperaturmessbereich
0.0-99.0 °C
Akkuleistung
4 × 1.5 V, 200 hr life
Allgemeine Beschreibung
With durable IP67 waterproof ABS housing, protective sensor cap, and an automatic shutdown feature that preserves battery life, OHAUS pen meters can endure consistent use in rough, wet environments.
Anwendung
pH, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO), salinity, total dissolved solids (TDS), temperature measurement
Leistungsmerkmale und Vorteile
Easily replaceable electrodes, automatic shut off feature
Rechtliche Hinweise
Ohaus is a registered trademark of Ohaus Corp.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)