Wichtige Dokumente
GBL722430
Grace Bio-Labs HybriSlip™ hybridization cover
L × W × thickness 24 mm × 30 mm × 0.25 mm, pkg of 1000 ea
Synonym(e):
coverslide, hydrophobic coverslip, nuclease free coverslip, thin plastic coverslip
About This Item
Empfohlene Produkte
Materialien
colorless
polycarbonate
Verpackung
pkg of 1000 ea
Hersteller/Markenname
Grace Bio-Labs 722430
L × B × Dicke
24 mm × 30 mm × 0.25 mm
Verwandte Kategorien
Allgemeine Beschreibung
HybriSlips are flexible, thin (0.25 mm) covers that minimize fluid friction and facilitate uniform reagent distribution. RNase free, hydrophobic covers will not trap or bind probes to their surfaces like coverglass. Product is ready-to-use without pretreatment. Working surfaces are protected with clean release liners to prevent RNase contamination. HybriSlips remain flat and will not curl, even at high temperatures.
- RNase and DNase free
- Hydrophobic
- Disposable
- Will not chip or break
Rechtliche Hinweise
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
GBL722430-1000EA | 4061832401300 |