Merck
EP0030108418
Eppendorf® DNA-LoBind-Röhrchen
capacity 1.5 mL, PCR clean, withstand, 30,000 x g, pack of 500 ea
Synonym(e):
Eppendorf® DNA LoBind tubes
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.15
Materialien
cap (push fit)
colorless
conical bottom
Beschreibung
Low retention
Sterilität
non-sterile
Leistungsmerkmale
DNase free
PCR clean
Verpackung
pack of 500 ea
Kapazität
1.5 mL
Eignung
suitable for PCR
Allgemeine Beschreibung
Maximize nucleic acid recovery. Eppendorf LoBind Tubes signifcantly reduce sample-to-surface binding to ensure maximal recovery of DNA and RNA molecules. The ideal solution for sample preparation and long-term storage of your precious nucleic acids
Leistungsmerkmale und Vorteile
Signifcantly reduce sample-to-surface binding to ensure maximal recovery of DNA and RNA molecules
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)