Merck
Z685100
Ace powder funnel with flat side
top diam. 75 mm, joint: ST/NS 14/20
About This Item
UNSPSC-Code:
41121809
NACRES:
NB.43
Materialien
borosilicate glass
Verbindung
joint: ST/NS14/20
Hersteller/Markenname
Ace Glass
Oberteil Durchm.
75 mm
Allgemeine Beschreibung
Ace powder funnel with standard taper joint and one flattened side for easy of use with multi-neck flasks.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)