Merck
Z685135
Ace powder funnel with flat side
top diam. 100 mm, joint: ST/NS 29/32
About This Item
UNSPSC-Code:
41121809
NACRES:
NB.43
Materialien
borosilicate glass
Verbindung
joint: ST/NS29/32
Hersteller/Markenname
Ace Glass
Oberteil Durchm.
100 mm
Allgemeine Beschreibung
Ace powder funnel with standard taper joint and one flattened side for easy of use with multi-neck flasks.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)