Merck
Z675431
Sterile economy sample spoon
1.25 mL
Synonym(e):
dispensing tool
About This Item
UNSPSC-Code:
41122403
NACRES:
NB.35
Beschreibung
PS
Verpackung
pack of 100 ea
Hersteller/Markenname
Bel-Art F36740-00001
Allgemeine Beschreibung
Moulded of lightweight and rigid, white high impact polystyrene. Individually packaged sterile spoons for one time use packed 100 per box.
  • Use to collect specific volumes of dry or liquid materials
  • Remains upright when filled or empty for use as a weighing boat
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)