Merck
Z657174
IKA® shaker attachment for HS/KS 260 basic and control shakers
AS 260.5 seperating funnel attachment, 1/cs
Synonym(e):
benchtop shakers, lab shakers, laboratory shakers
About This Item
UNSPSC-Code:
41103811
NACRES:
NB.73
Verpackung
case of 1
pack of 1
Hersteller/Markenname
IKA 3120900
Eignung
suitable for (for shaking out, salting out, extracting, eluting and sample enrichment)
Allgemeine Beschreibung
IKA® AS 260.5 Seperating Funnel Attachment
  • For shaking out, salting out, extracting, eluting, enriching.
  • The 3 clamping rolls (included in delivery) are height-adjustable for adaption to different separating funnel sizes.
  • The separating funnels are secured with O-rings (5 O-rings included).
Rechtliche Hinweise
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Kunden haben sich ebenfalls angesehen
Slide 1 of 1
1 of 1
Global Trade Item Number
SKUGTIN
Z657174-1EA4061832774329