Z645753
Chemical spill kit bag
Synonym(e):
chemical cleanup, spill cleanup, spill safety
Alle Fotos(1)
About This Item
UNSPSC-Code:
24111500
NACRES:
NB.35
Empfohlene Produkte
Hersteller/Markenname
Clear-spill CK3
Verfügbarkeit
not available in ROW, USA and Canada
Allgemeine Beschreibung
Kit designed for small to medium spills, ideal for laboratories or stores. Contains 40 pads, 4 x 8 cm by 1.2 m socks, 1 x 8 cm by 3 m sock, 1 x 30 cm by 35 cm cushion, 1 x 35 cm by 45 cm cushion with instructions supplied in a waterproof zip top bag with carry handles. Kit will absorb 57 litres.
PLEASE NOTE: KIT NO LONGER CONTAINS PPE
PLEASE NOTE: KIT NO LONGER CONTAINS PPE
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Kunden haben sich ebenfalls angesehen
Global Trade Item Number
SKU | GTIN |
---|---|
Z645753-1KT | 4061833446102 |