Merck
TMO6370-0004
Biohazard waste container
volume 5.5 L, polypropylene, pack of 1 ea
Synonym(e):
autoclave waste disposal, biohaz waste bag, biohazardous disposal bags, waste bags
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.35
Materialien
polypropylene
Verpackung
pack of 1 ea
Hersteller/Markenname
Nalge-Nunc 6370-0004
Volumen
5.5 L
5500 mL
Allgemeine Beschreibung
Biohazardous Waste Containers are ideal autoclavable polypropylene containers for secondary containment of biohazardous waste materials.
Convenient one-step sterilization; no need to remove bag. Port opening in cover allows one-handed disposal of materials; limits exposure to infectious waste. Holds standard sized autoclave bags. Will not rust, dent or puncture.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)