Merck
Z562521
LabJaws end-to-end lattice rod connector
jaw capacity 0-13 mm , 0-51 inches, aluminum
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.35
Materialien
aluminum
Hersteller/Markenname
Troemner 916133
Backenkapazität
0-13 mm , 0-51 inches
Allgemeine Beschreibung
Corrosion resistant. Increases the length of lab frame rods and clamps with perfect alignment. Comes with 2 set screws.
Rechtliche Hinweise
LabJaws is a trademark of Henry Troemner LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)