Z562505
LabJaws™ swivel clamp holder
jaw capacity 0-19 mm (0-0.75 in.), nickel plated (zinc)
Alle Fotos(1)
About This Item
UNSPSC-Code:
41122413
NACRES:
NB.35
Empfohlene Produkte
Materialien
nickel plated (zinc)
Hersteller/Markenname
Troemner 916065
Backenkapazität
0-19 mm (0-0.75 in.)
Allgemeine Beschreibung
Corrosion resistant. Has precise fit and powerful holding ability. Designed to enable tilting of clamp at any angle in parallel planes. Outside adjustment screw permits close distance between items being held.
Rechtliche Hinweise
LabJaws is a trademark of Henry Troemner LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Kunden haben sich ebenfalls angesehen
Global Trade Item Number
SKU | GTIN |
---|---|
Z562505-1EA | 4061833043042 |