Merck
Z301132
Quickfit® Erweiterungsadapter
top female joint: ST/NS 29/32, bottom male joint: ST/NS 19/26
Synonym(e):
XA42
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.35
Verbindung
female joint: ST/NS top29/32
male joint: ST/NS bottom19/26
Hersteller/Markenname
Quickfit XA 42
Verfügbarkeit
not available in USA and Canada
not available in USA and Canada
Allgemeine Beschreibung
These adapters connect items with larger cone to a piece of glassware with a smaller size socket.
Rechtliche Hinweise
Quickfit is a registered trademark of SciLabware Limited
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)