Merck
WHA77002808
Whatman® UNIFILTER® plates
size 96 wells, well working volume 800 μL, 0.45 μm cellulose acetate membrane
Synonym(e):
Z746487, micro plate, multiwell plate, well plates
About This Item
UNSPSC-Code:
41104900
NACRES:
NB.22
Materialien
clear polystyrene
conical bottom clear wells
polystyrene filter
Sterilität
non-sterile
Leistungsmerkmale
0.45 μm cellulose acetate membrane
Verpackung
pkg of 25 ea
Hersteller/Markenname
Whatman 7700-2808
Whatman Article No. 28420810 (US reference)
Maximales Volumen
800 μL
Größe
96 wells
Well-Arbeitsvolumen
800 μL
Porengröße
0.45 μm
Bindungsart
non-treated surface
Verwandte Kategorien
Allgemeine Beschreibung
  • Design prevents well to well crosstalk
  • Large variety of well volumes economizes on expensive consumables
  • Choice of filter media allows control of flow rates and retention characteristics
  • Standard overall dimensions fit most plate readers, auto samplers, robots
Anwendung
The UNIFILTER products are convenient, ready to use, disposable filter bottom microplates. Available in 96 well formats, the UNIFILTER microplates offer a choice of filter media that work well with the most stringent applications. UNIFILTERS can be used to process 96 samples at a time. The unique drip director design allows for analysis of the filtrate or the retentate. UNIFILTERS are available in many different variations of well formats and range from 100 μL to 2 mL, using a wide variety of filtration media.
Sonstige Hinweise
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Rechtliche Hinweise
UNIFILTER is a registered trademark of Cytiva
Whatman is a registered trademark of Cytiva
Die Dokumentenbibliothek aufrufen
Marit van Lenthe et al.
Malaria journal, 18(1), 430-430 (2019-12-20)
Sulfadoxine-pyrimethamine (SP) is a cornerstone of malaria chemoprophylaxis and is considered for programmes in the Democratic Republic of Congo (DRC). However, SP efficacy is threatened by drug resistance, that is conferred by mutations in the dhfr and dhps genes. The