Merck
TEM-CFT300CUUL
Sigma-Aldrich
Ultrathin Carbon Film Supported Copper Thin Bar Square Mesh Grid
size 300 mesh, box of 50
Synonym(e):
CARBON THIN BAR SQUARE MESH - CU 300 MESH UL 50/BX
About This Item
UNSPSC-Code:
41111761
NACRES:
MA.24
Verpackung
box of 50
Größe
300 mesh
Allgemeine Beschreibung
A 3-4 nm ultrathin film of pure carbon supported thin bar square mesh grid. Copper grid, 300 mesh. Carbon film show better electrical and heat conduction in the electron beam.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)