Merck
TEM-CF100HCU50
Sigma-Aldrich
Carbon Film Supported Copper Hexagonal Mesh Grid
size 100 mesh, box of 50
Synonym(e):
CARBON FILM ONLY ON 100 HEX COPPER GRIDS 50/BX
About This Item
UNSPSC-Code:
41111761
Materialien
carbon film
copper grid
Beschreibung
film layer thickness 5 - 6 nm carbon
Verpackung
box of 50
Größe
100 mesh
Allgemeine Beschreibung
A 5-6 nm thin film of pure carbon supported hexagonal mesh grids. Copper grid, 100 mesh. Carbon film show better electrical and heat conduction in the electron beam.