Merck
TEM-CF150CU50
Sigma-Aldrich
Carbon Film Supported Copper Grid
Standard Thickness, size 150 mesh, box of 50
Synonym(e):
Carbon Film 150 Mesh, Copper
About This Item
UNSPSC-Code:
41111761
NACRES:
MA.24
Materialien
carbon film
copper grid
Verpackung
box of 50
film size
5-6 nm
Größe
150 mesh
Allgemeine Beschreibung
TEM grids supported by carbon film only. A 5-6 nm thin film of pure carbon deposited on one side of the standard square mesh grid.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)