Merck
TEM-LC305CU
Sigma-Aldrich
Lacey Carbon Supported Copper Grids
hole size 50 nm, box of 5
Synonym(e):
LACEY CARBON FIL - CU 300 MESH 100 MICRON 5/BX
About This Item
UNSPSC-Code:
41111761
NACRES:
MA.24
Verpackung
box of 5
Lochgröße
50 nm
Allgemeine Beschreibung
Lacey carbon film supported TEM grids. This carbon coating consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)