Merck
SYNF611410
Synthware pressure equalizing addition funnel with metering stopcock
10 mL, joint: ST/NS 14/20
Synonym(e):
Synthware F611410, dropping funnel
About This Item
UNSPSC-Code:
41121809
NACRES:
NB.51
Verbindung
joint: ST/NS14/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F611410
Kapazität
10 mL
10 mL
Allgemeine Beschreibung
With PTFE adjustable bore metering plug. Turning the adjustment knob allows flow rate to be controlled from full flow to drop wise. The metering plug completely solves the problems of sudden overflow or non-flow during addition of reagents.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)