SYNF614125
Synthware™ pressure equalizing addition funnel with metering stopcock
125 mL, joint: ST/NS 24/40
Synonym(e):
Synthware F614125, dropping funnel
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121809
NACRES:
NB.51
Empfohlene Produkte
Verbindung
joint: ST/NS24/40
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F614125
Kapazität
125 mL
125 mL
Allgemeine Beschreibung
With PTFE adjustable bore metering plug. Turning the adjustment knob allows flow rate to be controlled from full flow to drop wise. The metering plug completely solves the problems of sudden overflow or non-flow during addition of reagents.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)