Merck
SYNA652440
Synthware long stem inlet adapter
joint: ST/NS 24/40
Synonym(e):
Synthware A652440, glass inlet adapter
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.51
Verbindung
joint: ST/NS24/40
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A652440
Allgemeine Beschreibung
With standard taper top outer and lower inner joints with sealed in lower tube extending below inner joint.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)