SYND381412
Synthware™ distilling receiver with 12 mL receiving tubes
joint: ST/NS 14/20
Synonym(e):
Synthware D381412, multi-tube distillation receiver
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.51
Empfohlene Produkte
Verbindung
joint: ST/NS14/20
Verpackung
pack of 1 ea
Hersteller/Markenname
D381412
Verwandte Kategorien
Allgemeine Beschreibung
Synthware™ distillation receiver features a standard taper joint (14/20) and three 12 mL receiving tubes graduated connected by three plastic joint clips with a 135° angle between the top outer joint and the lower inner joint.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)