Merck
HAM203230
Hamilton® SaltLine reagent syringe
model, 1750 TLL-SAL, no needle, volume 500 μL
About This Item
UNSPSC-Code:
42142600
NACRES:
NB.03
Materialien
Gastight syringe
Leistungsmerkmale
autoclavable: no
model 1750 TLL-SAL, no needle
Hersteller/Markenname
Hamilton
Parameter
20-50 °C temp. range (68-122 °F)
700 psig pressure
Volumen
500 μL
Verwandte Kategorien
Allgemeine Beschreibung
The SaltLine syringes are utilized in applications with high salt concentrations. They have the same PTFE Luer Lock termination but the plunger tip is made from Ultra High Molecular Weight Poly Ethylene (UHMWPE). The UHMWPE plunger tip resists damage that can be caused when salt crystal precipitate from the salt solution over time.
Rechtliche Hinweise
Hamilton is a registered trademark of Hamilton Co.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)