Merck
CLS1760125
Pyrex® tall form gas washing bottle, with ST stopper
capacity 125 mL
Synonym(e):
glass bottle
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Materialien
borosilicate glass bottle
clear bottle
cylindrical bottle
narrow-mouth bottle
Leistungsmerkmale
cap
Verpackung
case of 1 ea
pack of 1 ea
Hersteller/Markenname
Corning 1760-125
Kapazität
125 mL
Durchm. × H
41 mm × 245 mm
Rechtliche Hinweise
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)