BR91268
BRAND® glass beaker with spout, low form
volume 3000 mL, with graduation
Synonym(e):
Beakers, Glass
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121803
NACRES:
NB.43
Empfohlene Produkte
Materialien
borosilicate glass 3.3
Leistungsmerkmale
with graduation
Hersteller/Markenname
BRAND 91268
Volumen
3000 mL
Verwandte Kategorien
Allgemeine Beschreibung
The BRAND® Glass Beaker with Spout is expertly crafted from Boro 3.3 glass, making it a reliable choice for laboratory applications. Designed in accordance with DIN EN ISO 3819 standards, this low-form beaker features clear graduation markings for precise measurement of liquids. The integrated spout allows for easy pouring, minimizing spills and enhancing accuracy during liquid transfer.
Sonstige Hinweise
Browse all glass beakers
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR91268-4EA | 4061831867831 |