BR768505
BRAND® tidy tray with compartments
5 compartments
Synonym(e):
Trays
Alle Fotos(1)
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.51
Empfohlene Produkte
Materialien
PVC
Verpackung
pack of 1 ea
Hersteller/Markenname
BRAND 768505
Größe
400 mm × 300 mm × 65 mm
Allgemeine Beschreibung
The BRAND® tidy tray with compartments is made from durable PVC and is specifically designed for use as a utility tray or drawer organizer. Its compartmentalized structure allows for efficient storage and organization of various laboratory items, such as magnetic stirring bars and other small tools.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR768505-1EA | 4061831859348 |