Merck
254088
Sigma-Aldrich
Gold(I)-cyanid
99.9% trace metals basis
About This Item
Lineare Formel:
AuCN
CAS-Nummer:
Molekulargewicht:
222.98
EG-Nummer:
MDL-Nummer:
UNSPSC-Code:
12352300
PubChem Substanz-ID:
NACRES:
NA.23
Qualitätsniveau
Assay
99.9% trace metals basis
Form
powder
Eignung der Reaktion
core: gold
Dichte
7.14 g/mL at 25 °C (lit.)
SMILES String
[Au]C#N
InChI
1S/CN.Au/c1-2;
InChIKey
DRJJJWGHLJZDGQ-UHFFFAOYSA-N
Piktogramme
Skull and crossbonesEnvironment
Signalwort
Danger
Gefahreneinstufungen
Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1
Zusätzliche Gefahrenhinweise
Lagerklassenschlüssel
6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Persönliche Schutzausrüstung
dust mask type N95 (US), Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Kunden haben sich ebenfalls angesehen
Philip M Yangyuoru et al.
Journal of inorganic biochemistry, 102(3), 576-583 (2008-02-08)
Electrospray ionization spectra of potential cyanide-containing gold-drug metabolites revealed additional, weak, unanticipated peaks at approximately twice the mass of the gold(I) and gold(III) cyanide complexes. The exact masses correspond to proton-linked bimetallic complexes, [H[Au(CN)(m)](2)](-), (m=2,4). Further investigation revealed a total
G G Graham et al.
Biochemical pharmacology, 56(3), 307-312 (1998-09-23)
There is considerable evidence that the anti-rheumatic gold complexes are activated by their conversion to aurocyanide. In order to understand the mechanism of production of aurocyanide, we investigated the involvement of myeloperoxidase in the reaction. This haem enzyme of neutrophils
Hong-ping Tang et al.
Guang pu xue yu guang pu fen xi = Guang pu, 24(6), 752-755 (2005-03-16)
Quaternary ammonium salt has strong affinity with gold in cyanide solutions, but it is not readily stripped. Stripping with acidic thiourea solutions and air sparging shows promise. There are some reports on the process of thiourea stripping, but little about
Xiandi Gong et al.
The Journal of general physiology, 122(6), 673-687 (2003-11-12)
Chloride permeation through the cystic fibrosis transmembrane conductance regulator (CFTR) Cl- channel is blocked by a broad range of anions that bind tightly within the pore. Here we show that the divalent anion Pt(NO2)42- acts as an impermeant voltage-dependent blocker
Xiandi Gong et al.
The Journal of physiology, 540(Pt 1), 39-47 (2002-04-03)
Lyotropic anions with low free energy of hydration show both high permeability and tight binding in the cystic fibrosis transmembrane conductance regulator (CFTR) Cl(-) channel pore. However, the molecular bases of anion selectivity and anion binding within the CFTR pore