MilliporeSigma
All Photos(2)
Key Documents
MAK014
Sigma-Aldrich
Glucose-6-Phosphate Assay Kit
sufficient for 100 colorimetric tests
Sign Into View Organizational & Contract Pricing
About This Item
UNSPSC Code:
12161503
NACRES:
NA.84
usage
sufficient for 100 colorimetric tests
detection method
colorimetric
relevant disease(s)
hematological disorder
storage temp.
−20°C
General description
Glucose 6-phosphate (G6P) is a key metabolic intermediate that enters either metabolic pathways or storage. G6P is generated when glucose is phosphorylated by hexokinase or glucokinase or by the conversion of glucose-1-phosphate by phosphoglucomutase during glycogenolysis. G6P lies at the beginning of both glycolysis and the pentose phosphate pathways. It also can be stored as glycogen when blood glucose levels are high.
Application
Glucose-6-phosphate assay kit has been used in:
  • glucose 6-phosphate (G6P) assay
  • quantification of glucose-6-phosphate (G6P) and fructose-6-phosphate (F6P)
  • metabolite analysis
Suitability
Suitable for quantifying Glucose-6-Phosphate in a variety of samples.
Principle
G6P is utilized by Glucose-6-phosphate dehydrogenase to generate reducing equivalents in the form of NADPH. This is particularly important in red blood cells where a G6PDH deficiency leads to hemolytic anemia. G6P is determined by an enzyme assay, which results in a colorimetric (450 nm) product, proportional to the G6P present. Typical sensitivities of detection are between 1-30 nmoles.
replaced by
pictograms
Health hazardCorrosion
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B
Storage Class
8A - Combustible corrosive hazardous materials
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Renal Disease in Type I Glycogen Storage Disease
Richard P L, et al.
Genetic Diseases of the Kidney (2009)
Reciprocal Control of Anaplerotic Phosphoenolpyruvate Carboxylase by in Vivo Monoubiquitination and Phosphorylation in Developing Proteoid Roots of Phosphate-Deficient Harsh Hakea
Shane M W, et al.
Plant Physiology, 112-112 (2013)
MiR-182 promotes glucose metabolism by upregulating hypoxia-inducible factor 1a in NSCLC cells
Mingliang W, et al.
Biochemical and Biophysical Research Communications, 504(2), 400-405 (2018)
Saixiang Feng et al.
Veterinary research, 51(1), 97-97 (2020-08-02)
Lipooligosaccharides (LOSs) are virulence determinants of Glaesserella parasuis, a pathogen of the respiratory tract of pigs. We previously reported that disruption of the galU or galE gene in G. parasuis results in increased sensitivity to porcine serum, indicating that the
Forkhead Box O6 (FoxO6) Depletion Attenuates Hepatic Gluconeogenesis and Protects against Fat-induced Glucose Disorder in Mice.
Virtu C N, et al.
The Journal of Biological Chemistry, 290, 15581-15594 (2015)
Articles
We presents an article about the Warburg effect, and how it is the enhanced conversion of glucose to lactate observed in tumor cells, even in the presence of normal levels of oxygen. Otto Heinrich Warburg demonstrated in 1924 that cancer cells show an increased dependence on glycolysis to meet their energy needs, regardless of whether they were well-oxygenated or not.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service