Ligation of Insert to pGEX DNA
Ready-To-Go T4 DNA Ligase can be used to achieve ligations in 30 to 45 min. An alternate procedure is described below.
Reagents Required |
---|
Procedure
- Prepare linearized pGEX DNA and insert DNA so that they will be present at a vector to insert ratio of 1:5 moles of ends. The moles of ends of linear DNA can be calculated with the following formula:
moles of ends = 2 × (g of DNA)/[(# of bp) × (649 Daltons/bp)]
Example: 100 ng of pGEX DNA (0.06 pmol of ends) would require 100 ng of a 1 kb insert (0.3 pmol of ends).
- For ligation of cohesive ends, the final reaction mix should contain 1 mM ATP (diluted) and 0.5 to 5 units of T4 DNA ligase, and should be incubated for 1 to 4 h at 10 °C
- For ligation of blunt ends, the final reaction mix should contain 0.1 to 1 mM ATP (diluted) and 10 to 15 units of T4 DNA ligase, and should be incubated for 2 to 16 h at 4 °C to 16 °C.
- Based upon the above considerations in step 1, prepare the following reaction mixture specific for your application:
1 to 5 µl of linearized pGEX DNA
1 to 5 µL of insert DNA
2 µL of 10× One-Phor-All Buffer PLUS (OPA+)
0.2 µL of 100 mM ATP
0.5 to 15 units of T4 DNA ligase
Water to 20 µL
- Incubate for either 1 to 4 h at 10 °C (cohesive ends) or 2 to 16 h at 4 °C to 16 °C (blunt ends).
- Terminate the reaction by heating at 65 °C for 10 min.
The ligation reaction can be used directly to transform competent cells. Otherwise, it can be stored at -20 °C until needed.