GC Analysis of Class 3 Residual Solvents on SUPELCOWAX 10
Materiali
Colonna analitica
N° Catalogo
Descrizione
Determinazione del prezzo
CONDITIONS
column
SUPELCOWAX 10, 30 m x 0.53 mm I.D., 1.0 μm (25301-U)
oven
35 °C (15 min), 5 °C/min to 200 °C
inj. temp.
225 °C
detector
FID, 250 °C
carrier gas
helium, 30 cm/sec @ 35 °C
injection
1.0 μL 33:1 split
liner
single taper
sample
27 analytes, each at 3000 μg/mL in methanol
Descrizione
Note legali
null