The Milli-Q® IQ 7003/7005/7010/7015 fully-integrated water purification system is your ultimate lab water solution, providing superior quality Type 1 and Type 2 water directly from tap water. With Elix® EDI technology, intuitive touchscreens, Q-POD® and E-POD® dispensers, and mercury-free UV lamps, the system will exceed your most demanding expectations – from improved productivity and versatility, to reduced environmental impact.
Milli-Q® IQ 7003/05/10/15
Features & Benefits
  • Simple & intuitive dispensing – You’ll always have convenient and easy access to purified water with our ergonomic Q-POD® ultrapure and E-POD® pure water dispensers. Just twist the dispense wheel or tap the touchscreen menu to access a range of dispensing functions.
  • Consistently superior quality water – Our exclusive, advanced and sustainable purification technologies—including mercury-free ech2o® UV lamps, Elix® EDI technology and IQnano® ion-exchange resin—deliver highest-quality Type 1 and Type 2 water for your critical experiments. Our range of POD-Paks lets you further match water quality to your every application. And with accurate, rapid and continuous resistivity and TOC monitoring directly on the POD touchscreens, you’ll always have full confidence in your water quality.
  • Assured protection of stored water – Our redesigned intelligent pure water storage solution ensures that high-quality Type 2 water is always on hand and ready to use. Unique design features protect against stagnation and bacterial contamination, safeguarding the quality of stored pure water better than ever before.
  • Unparalleled ease of use – Choose a dispense option, instantly monitor water quality parameters, or rapidly retrieve data. Easy-to-navigate touchscreens on each POD dispenser give you quick access and control of essential system functions in just a few clicks.
  • Space-saving & clutter-free – Save valuable space as only the slim POD dispenser stays on the bench. The compact purification unit and tank can be tucked away under the bench, on the wall, or a combination of both to best fit your lab’s space. A range of production flow rates (3,5,10,15 L/h) and tank size (25,50,100L) options are available to ensure your lab's needs can be met today and tomorrow. For labs needing more than one point-of-use, up to 4 dispensers can be connected to one system (1 E-POD® and 3 Q-POD® units, or 4 Q-POD® units).
  • Effortless data management – Simplified data management is an integral part of the Milli-Q® IQ 7003/05/10/15 system. In a few clicks, download an individual Dispense Report, determine water quality over a specified time frame, allocate costs for shared resources, or create a customized report. Traceability is also ensured through the e-Sure tags on all consumables.
  • Completely mercury free – The Milli-Q® IQ 7003/05/10/15 system is the first and only mercury-free tap to ultrapure water system, with patented built-in mercury-free ech2o® UV lamps. Additionally, advanced RO purification, with its RO recovery loop, optimizes water recovery to reduce water consumption. Plus “LabClose” mode puts the system into hibernation, to save energy while preserving the system water quality when you are away from the lab.
  • Quality & Compliance – Validated and qualified design, combined with our strict manufacturing Quality Assurance system, is your best warranty. We offer a complete range of services, including validation support, which is provided by certified field service engineers using calibrated equipment and a Milli-Q®-specific Qualification Workbook. A Compliance Report on Milli-Q® IQ 7003/05/10/15 system water conformity to industry norms (including ASTM®, ISO® 3696, CLSI®, USP and EP) is available upon request.
Water Applications
Ultrapure Water Applications
  • Chromatography: HPLC, LC-MS, UHPLC, ion chromatography (IC)
  • Elemental analyses: AAS, ICP-MS, ICP-OES
  • Life science applications: Cell culture, PCR, sequencing, DNA microarrays, protein analyses
Pure Water Applications
  • Analytical techniques with sensitivity at ppm level or above, such as pH measurements, UV spectrophotometry, Kjeldahl testing
  • Reagent preparation for dissolution testing, chemical synthesis, microbiology media preparation
  • Staining solutions preparation for histology
  • Feed for instruments: glassware washers, stainless steel autoclaves, stability and weathering test chambers, clinical analyzers, hydrogen generators
Water Specifications / International Conformity
1 These values are typical and may vary depending on the nature and concentration of contaminants in the feed water.
2 Resistivity can also be displayed non-temperature-compensated as required by USP.
3 In the appropriate operating conditions, otherwise typically ≤ 5 ppb.
4 With Millipak® filter or Millipak® Gold filter.
5 With Millipak® or Biopak® filter when installed and used in a laminar flow hood.
6 With Biopak® polisher when installed and used in a laminar flow hood.
7 With Biopak® polisher
Feed Water Requirements
International Regulation Requirements
Declaration of CE and cUL Conformity:
The Milli-Q® IQ 7003, Milli-Q® IQ 7005, Milli-Q® IQ 7010 and Milli-Q® IQ 7015 systems have been tested by an independent and accredited company for compliance with CE directives related to safety and electromagnetic compatibility. The report can be consulted on demand. In addition, the Milli-Q® IQ 7003/05/10/15 systems are built using components and practices recommended by UL and has been cUL marked. The registration can be verified on the UL web site: www.ul.com.
Video: Superior quality Type 1 and Type 2 water from tap
See for yourself how the Milli-Q® IQ 7003/05/10/15 ultrapure and pure water purification system is designed to exceed your most demanding expectations – from improved productivity and versatility, to reduced environmental impact.
Video: Customer Testimonial