MilliporeSigma
  • Differential Expression of miR-93 and miR-21 in Granulosa Cells and Follicular Fluid of Polycystic Ovary Syndrome Associating with Different Phenotypes.
Differential Expression of miR-93 and miR-21 in Granulosa Cells and Follicular Fluid of Polycystic Ovary Syndrome Associating with Different Phenotypes.
Scientific reports (2017-11-09)
Mohammad Naji, Ashraf Aleyasin, Saeid Nekoonam, Ehsan Arefian, Reza Mahdian, Fardin Amidi
ABSTRACT
The heterogeneous and multifactorial essence of polycystic ovary syndrome (PCOS) renders a remarkable significance to microRNAs (miRNAs). Normo-androgenic (NA) and hyperandrogenic (HA) PCOS patients were compared with matched healthy women. Expression of miRNAs and TGFβ signaling genes was studied by qRT-PCR and western blotting. Effect of androgen on expression of miR-93 and miR-21 and involvement of androgen receptor were appraised. In granulosa cells (GCs), miR-93 and miR-21 showed significantly increased levels in HA patients compared to NA patients. On the contrary, follicular fluid (FF) levels of both miRNAs were significantly decreased in HA group compared to control women. No significant change in the expression of miRNAs in serum samples was detected. Furthermore, mRNA levels of SMAD7 and TGFBR2 were significantly downregulated in GCs of HA group compared to NA and control subjects. TGFBR2 protein level was significantly decreased in HA patients compared to controls. Free testosterone and free androgen index were positively correlated with expression of miR-93 and miR-21 in GCs of PCOS group. Our findings show distinct molecular signature of different subtypes of PCOS. Intermediary position of miRNAs as androgen responsive factors may play critical role in the pathogenesis of PCOS in hyperandrogenic condition.