MilliporeSigma
  • Maternal intake of grape seed procyanidins during lactation induces insulin resistance and an adiponectin resistance-like phenotype in rat offspring.
Maternal intake of grape seed procyanidins during lactation induces insulin resistance and an adiponectin resistance-like phenotype in rat offspring.
Scientific reports (2017-10-05)
Antoni Caimari, Roger Mariné-Casadó, Noemí Boqué, Anna Crescenti, Lluís Arola, Josep Maria Del Bas
ABSTRACT
Previously, we demonstrated that a grape seed procyanidin extract (GSPE) supplementation in pregnant and lactating rats exerted both healthy and deleterious programming effects on their offspring. Here, we evaluated whether the administration of GSPE during lactation (100 mg.kg