MilliporeSigma
Transport characteristics of urea transporter-B.
Sub-cellular biochemistry (2014-10-10)
Baoxue Yang
ABSTRACT
UT-B represents the major urea transporter in erythrocytes, in addition to being expressed in kidney descending vasa recta, brain, spleen, ureter, bladder, and testis. Expression of urea transporter UT-B confers high urea permeability to mammalian erythrocytes. Erythrocyte membranes are also permeable to various urea analogues, suggesting common transport pathways for urea and structurally similar solutes. UT-B is highly permeable to urea and the chemical analogues formamide, acetamide, methylurea, methylformamide, ammonium carbamate, and acrylamide, each with a Ps > 5.0 × 10(-6) cm/s at 10 °C. The amides formamide, acetamide, acrylamide, and butyramide efficiently diffuse across lipid bilayers. The urea analogues dimethylurea, acryalmide, methylurea, thiourea, and methylformamide inhibit UT-B-mediated urea transport by >60 % by a pore-blocking mechanism. UT-B is also a water channel in erythrocytes and has a single-channel water permeability that is similar to aquaporin-1. Whether UT-B is an NH3 channel still needs further study. Urea permeability (Purea) in erythrocytes differs between different mammals. Carnivores (dog, fox, cat) exhibit high Purea. In contrast, herbivores (cow, donkey, sheep) show much lower Purea. Erythrocyte Purea in human and pig (omnivores) was intermediate. Rodents and lagomorphs (mouse, rat, rabbit) have Purea intermediate between carnivores and omnivores. Birds that do not excrete urea and do not express UT-B in their erythrocytes have very low values. In contrast to Purea, water permeability is relatively similar in all mammals studied. This chapter will provide information about the transporter characteristics of UT-B.