MilliporeSigma
  • The introduction of RNA-DNA differences underlies interindividual variation in the human IL12RB1 mRNA repertoire.
The introduction of RNA-DNA differences underlies interindividual variation in the human IL12RB1 mRNA repertoire.
Proceedings of the National Academy of Sciences of the United States of America (2015-12-02)
Amy J Turner, Praful Aggarwal, Halli E Miller, Jill Waukau, John M Routes, Ulrich Broeckel, Richard T Robinson
ABSTRACT
Human interleukin 12 and interleukin 23 (IL12/23) influence susceptibility or resistance to multiple diseases. However, the reasons underlying individual differences in IL12/23 sensitivity remain poorly understood. Here we report that in human peripheral blood mononuclear cells (PBMCs) and inflamed lungs, the majority of interleukin-12 receptor β1 (IL12RB1) mRNAs contain a number of RNA-DNA differences (RDDs) that concentrate in sequences essential to IL12Rβ1's binding of IL12p40, the protein subunit common to both IL-12 and IL-23. IL12RB1 RDDs comprise multiple RDD types and are detectable by next-generation sequencing and classic Sanger sequencing. As a consequence of these RDDs, the resulting IL12Rβ1 proteins have an altered amino acid sequence that could not be predicted on the basis of genomic DNA sequencing alone. Importantly, the introduction of RDDs into IL12RB1 mRNAs negatively regulates IL12Rβ1's binding of IL12p40 and is sensitive to activation. Collectively, these results suggest that the introduction of RDDs into an individual's IL12RB1 mRNA repertoire is a novel determinant of IL12/23 sensitivity.