MilliporeSigma
  • The interaction between HIV-1 Nef and adaptor protein-2 reduces Nef-mediated CD4
The interaction between HIV-1 Nef and adaptor protein-2 reduces Nef-mediated CD4
Virology (2017-06-04)
Rajesh Abraham Jacob, Aaron L Johnson, Emily N Pawlak, Brennan S Dirk, Logan R Van Nynatten, S M Mansour Haeryfar, Jimmy D Dikeakos
ABSTRACT
Acquired Immune Deficiency Syndrome is characterized by a decline in CD4