MilliporeSigma
  • SRC family kinases in hamster spermatozoa: evidence for the presence of LCK.
SRC family kinases in hamster spermatozoa: evidence for the presence of LCK.
Reproduction (Cambridge, England) (2017-03-03)
Durgesh Kumar Singh, Rohit Kumar Deshmukh, Praveen Kumar Narayanan, Sisinthy Shivaji, Archana Bharadwaj Siva
ABSTRACT
Sperm capacitation is a prerequisite for successful fertilization. Increase in tyrosine phosphorylation is considered the hallmark of capacitation and attempts to understand its regulation are ongoing. In this regard, we attempted to study the role of SRC family kinases (SFKs) in the hamster sperm functions. Interestingly, we found the presence of the lymphocyte-specific protein tyrosine kinase, LCK, in mammalian spermatozoa and further characterized it in terms of its localization and function. LCK was found in spermatozoa of several species, and its transcript was identified in the hamster testis. Autophosphorylation of LCK at the Y394 residue increased as capacitation progressed, indicating an upregulation of LCK activity during capacitation. Inhibition of LCK (and perhaps the other SFKs) with the use of a specific inhibitor showed a significant decrease in protein tyrosine phosphorylation of several proteins, implying LCK/SFKs as key tyrosine kinase(s) regulating tyrosine phosphorylation during hamster sperm capacitation. Dihydrolipoamide dehydrogenase was identified as a substrate for LCK/SFK. LCK/SFKs inhibition significantly reduced the percentage fertilization (