MilliporeSigma
  • Characterization of the Zika virus two-component NS2B-NS3 protease and structure-assisted identification of allosteric small-molecule antagonists.
Characterization of the Zika virus two-component NS2B-NS3 protease and structure-assisted identification of allosteric small-molecule antagonists.
Antiviral research (2017-05-04)
Sergey A Shiryaev, Chen Farhy, Antonella Pinto, Chun-Teng Huang, Nicole Simonetti, Annie Elong Ngono, Antimone Dewing, Sujan Shresta, Anthony B Pinkerton, Piotr Cieplak, Alex Y Strongin, Alexey V Terskikh
ABSTRACT
The recent re-emergence of Zika virus (ZIKV)