MilliporeSigma
Microbial fuel cells using Cellulomonas spp. with cellulose as fuel.
Journal of bioscience and bioengineering (2016-11-08)
Yuya Takeuchi, Wichean Khawdas, Yuji Aso, Hitomi Ohara
ABSTRACT
Cellulomonas fimi, Cellulomonas biazotea, and Cellulomonas flavigena are cellulose-degrading microorganisms chosen to compare the degradation of cellulose. C. fimi degraded 2.5 g/L of cellulose within 4 days, which was the highest quantity among the three microorganisms. The electric current generation by the microbial fuel cell (MFC) using the cellulose-containing medium with C. fimi was measured over 7 days. The medium in the MFC was sampled every 24 h to quantify the degradation of cellulose, and the results showed that the electric current increased with the degradation of cellulose. The maximum electric power generated by the MFC was 38.7 mW/m